ENA292596253 Enamine

2-amino-N-(6-methoxypyridin-3-yl)benzamide

  • MDL number MFCD09739779
  • Cas Number : 954259-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61786
Availability details for
Quantity
  • Availability
  • Add to Cart