ENA292596265 Enamine

N-cyclopropyl-1-(pyridin-4-yl)piperidin-4-amine

  • MDL number MFCD11106606
  • Cas Number : 1152836-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61811
Availability details for
Quantity
  • Availability
  • Add to Cart