ENA292596267 Enamine

2-amino-1-(naphthalen-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD11974305
  • Cas Number : 53773-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-250
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61814
Availability details for
Quantity
  • Availability
  • Add to Cart