ENA292596268 Enamine

2-hydroxy-5-phenylbenzaldehyde

  • MDL number MFCD00579079
  • Cas Number : 1761-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61815
Availability details for
Quantity
  • Availability
  • Add to Cart