ENA292596269 Enamine

1-(pyridin-4-ylmethyl)-1H-pyrazol-4-amine

  • MDL number MFCD11128186
  • Cas Number : 1152841-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61816
Availability details for
Quantity
  • Availability
  • Add to Cart