ENA292596270 Enamine

6-(2-methyl-1H-imidazol-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD09940807
  • Cas Number : 1016839-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61817
Availability details for
Quantity
  • Availability
  • Add to Cart