ENA292596271 Enamine

2-({[(3-nitrophenyl)methyl]carbamoyl}amino)acetic acid

  • MDL number MFCD09805900
  • Cas Number : 1016510-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61818
Availability details for
Quantity
  • Availability
  • Add to Cart