ENA292596273 Enamine

5-methyl-1-[2-(propan-2-yl)phenyl]-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106444
  • Cas Number : 1154372-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61820
Availability details for
Quantity
  • Availability
  • Add to Cart