ENA292596277 Enamine

2-[2-(furan-2-yl)-5-methyl-1,3-oxazol-4-yl]acetic acid

  • MDL number MFCD07379009
  • Cas Number : 929972-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61824
Availability details for
Quantity
  • Availability
  • Add to Cart