ENA292596279 Enamine

4-(1H-imidazol-2-yl)phenol

  • MDL number MFCD08668843
  • Cas Number : 15548-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265...267
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61826
Availability details for
Quantity
  • Availability
  • Add to Cart