ENA292596281 Enamine

1-(2,6-diethylphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD01938069
  • Cas Number : 25372-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61828
Availability details for
Quantity
  • Availability
  • Add to Cart