ENA292596287 Enamine

methyl 2-(2H-1,2,3,4-tetrazol-5-yl)acetate

  • MDL number MFCD02594444
  • Cas Number : 26476-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61836
Availability details for
Quantity
  • Availability
  • Add to Cart