ENA292596293 Enamine

2-(2-chloroethyl)thiophene

  • MDL number MFCD11646274
  • Cas Number : 19995-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61842
Availability details for
Quantity
  • Availability
  • Add to Cart