ENA292596294 Enamine

3-(3-hydroxypropyl)-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD12186729
  • Cas Number : 20844-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61844
Availability details for
Quantity
  • Availability
  • Add to Cart