ENA292596308 Enamine

4-(pyridin-4-ylsulfanyl)phenol

  • MDL number MFCD12824685
  • Cas Number : 64557-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61869
Availability details for
Quantity
  • Availability
  • Add to Cart