ENA292596315 Enamine

1-(6-phenoxypyridin-3-yl)methanamine

  • MDL number MFCD09738654
  • Cas Number : 620608-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61877
Availability details for
Quantity
  • Availability
  • Add to Cart