ENA292596331 Enamine

[2-(4-fluorophenoxy)pyridin-4-yl]methanamine

  • MDL number MFCD09741151
  • Cas Number : 869293-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61376

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61376
Availability details for
Quantity
  • Availability
  • Add to Cart