ENA292596332 Enamine

[6-(3-fluorophenoxy)pyridin-3-yl]methanamine

  • MDL number MFCD09735325
  • Cas Number : 953906-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61379
Availability details for
Quantity
  • Availability
  • Add to Cart