ENA292596335 Enamine

2-amino-2-(1-methyl-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD11155567
  • Cas Number : 195070-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61917

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61917
Availability details for
Quantity
  • Availability
  • Add to Cart