ENA292596337 Enamine

1-(1,3-dioxaindane-5-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09050078
  • Cas Number : 142962-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61919
Availability details for
Quantity
  • Availability
  • Add to Cart