ENA292596342 Enamine

2-amino-N-methoxy-N-methylbenzamide

  • MDL number MFCD12095390
  • Cas Number : 133776-41-7
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61925
Availability details for
Quantity
  • Availability
  • Add to Cart