ENA292596348 Enamine

4-[2-(5-phenyl-2H-1,2,3,4-tetrazol-2-yl)ethyl]aniline

  • MDL number MFCD12582962
  • Cas Number : 1155622-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61951
Availability details for
Quantity
  • Availability
  • Add to Cart