ENA292596358 Enamine

4-methanesulfonylbenzene-1-thiol

  • MDL number MFCD01911822
  • Cas Number : 3814-25-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-68
Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61963

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-61963
pictograms GHS06
hstatements H301,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart