ENA292596360 Enamine

2-(5-bromothiophene-2-sulfonamido)acetic acid

  • MDL number MFCD03834404
  • Cas Number : 706766-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61965
Availability details for
Quantity
  • Availability
  • Add to Cart