ENA292596367 Enamine

[6-(cyclohexyloxy)pyridin-3-yl]methanamine

  • MDL number MFCD09258826
  • Cas Number : 928648-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61973
Availability details for
Quantity
  • Availability
  • Add to Cart