ENA292596369 Enamine

4-(2-methylphenyl)pyrrolidin-2-one

  • MDL number MFCD03447336
  • Cas Number : 654633-87-1
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62004
Availability details for
Quantity
  • Availability
  • Add to Cart