ENA292596375 Enamine

N-(3-bromophenyl)piperidine-4-carboxamide

  • MDL number MFCD09805966
  • Cas Number : 1016503-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62011
Availability details for
Quantity
  • Availability
  • Add to Cart