ENA292596376 Enamine

1-(2-methoxyphenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD09864776
  • Cas Number : 74205-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62012

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62012
Availability details for
Quantity
  • Availability
  • Add to Cart