ENA292596384 Enamine

N-(4-bromophenyl)-1-methylpiperidin-4-amine

  • MDL number MFCD11144879
  • Cas Number : 93591-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62021
Availability details for
Quantity
  • Availability
  • Add to Cart