ENA292596385 Enamine

N-(2,2-dichloroethenyl)-4-fluorobenzamide

  • MDL number MFCD01409258
  • Cas Number : 67833-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61453
Availability details for
Quantity
  • Availability
  • Add to Cart