ENA292596391 Enamine

2-methyl-1-(pyridin-2-yl)propan-1-ol

  • MDL number MFCD12153509
  • Cas Number : 102439-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62031
Availability details for
Quantity
  • Availability
  • Add to Cart