ENA292596394 Enamine

3-[(3-methoxyphenyl)amino]pyrrolidine-2,5-dione

  • MDL number MFCD12099363
  • Cas Number : 1218258-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62034
Availability details for
Quantity
  • Availability
  • Add to Cart