ENA292596401 Enamine

3-(2-chloropropanamido)-N-methylbenzamide

  • MDL number MFCD11643907
  • Cas Number : 1098346-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28040
Availability details for
Quantity
  • Availability
  • Add to Cart