ENA292596410 Enamine

3-(3-bromophenyl)-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD12774154
  • Cas Number : 26904-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62063

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62063
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart