ENA292596413 Enamine

3-amino-N-(5-bromopyridin-2-yl)benzamide

  • MDL number MFCD11184844
  • Cas Number : 1039972-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62066

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62066
Availability details for
Quantity
  • Availability
  • Add to Cart