ENA292596414 Enamine

5-[(4-chloro-1H-pyrazol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD01852387
  • Cas Number : 312309-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62068

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62068
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart