ENA292596424 Enamine

N-[(5-bromo-2-fluorophenyl)methyl]cyclopropanamine

  • MDL number MFCD09047386
  • Cas Number : 926201-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33120
Availability details for
Quantity
  • Availability
  • Add to Cart