ENA292596425 Enamine

4-[(4-methoxyphenyl)methyl]piperidine

  • MDL number MFCD05863571
  • Cas Number : 37581-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-61
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62082
Availability details for
Quantity
  • Availability
  • Add to Cart