ENA292596426 Enamine

4-methoxynaphthalene-1-thiol

  • MDL number MFCD11204081
  • Cas Number : 91344-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62083
Availability details for
Quantity
  • Availability
  • Add to Cart