ENA292596428 Enamine

1-(1-hydroxynaphthalene-2-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09946224
  • Cas Number : 1103247-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62085
Availability details for
Quantity
  • Availability
  • Add to Cart