ENA292596432 Enamine

2-(2-methylphenyl)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD09812699
  • Cas Number : 152954-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62090

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62090
Availability details for
Quantity
  • Availability
  • Add to Cart