ENA292596434 Enamine

3,4-dichloro-N-(propan-2-yl)aniline

  • MDL number MFCD11143457
  • Cas Number : 54962-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62092
Availability details for
Quantity
  • Availability
  • Add to Cart