ENA292596436 Enamine

1-[(pyridin-2-yl)methyl]-1H-pyrazol-4-amine

  • MDL number MFCD11128206
  • Cas Number : 1152940-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62094
Availability details for
Quantity
  • Availability
  • Add to Cart