ENA292596437 Enamine

1-[(pyridin-3-yl)methyl]-1H-pyrazol-4-amine

  • MDL number MFCD11128207
  • Cas Number : 1152940-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62095
Availability details for
Quantity
  • Availability
  • Add to Cart