ENA292596439 Enamine

3-(cyclopentylmethyl)-1,2-oxazol-5-amine

  • MDL number MFCD10689548
  • Cas Number : 794566-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62106

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62106
Availability details for
Quantity
  • Availability
  • Add to Cart