ENA292596440 Enamine

3-(2-methoxyphenyl)-1,2-oxazol-5-amine

  • MDL number MFCD07383032
  • Cas Number : 54606-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61464
Availability details for
Quantity
  • Availability
  • Add to Cart