ENA292596442 Enamine

2,6-dichloropyridine-3-sulfonamide

  • MDL number MFCD15142801
  • Cas Number : 1078627-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62109
Availability details for
Quantity
  • Availability
  • Add to Cart