ENA292596446 Enamine

4-(2-chloroethyl)benzene-1-sulfonamide

  • MDL number MFCD00460202
  • Cas Number : 5378-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62113

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62113
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart