ENA292596447 Enamine

4-(1H-1,2,4-triazol-1-yl)-3-(trifluoromethyl)aniline

  • MDL number MFCD12768357
  • Cas Number : 351325-27-4
  • Molecular Weight : 228.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62114
Availability details for
Quantity
  • Availability
  • Add to Cart