ENA292596448 Enamine

3-(pyridin-3-yloxy)propanoic acid

  • MDL number MFCD09034760
  • Cas Number : 933711-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62115

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62115
Availability details for
Quantity
  • Availability
  • Add to Cart